Skip to Content
Merck
All Photos(1)

Key Documents

IAK0108

Sigma-Aldrich

Phagocytosis Assay Kit

Green Zymosan, sufficient for 100 fluorometric assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Phagocytosis is a cellular process for ingesting and eliminating microorganisms, foreign substances, and apoptotic cells. It is a specific form of endocytosis initiated by recognition and binding of foreign particles by cell surface receptors, followed by their engulfment, and formation of phagosomes. Maturing phagosomes transform to phagolysosomes which destroy the pathogen through enzymes and toxic peroxides. Zymosan prepared from yeast cell wall (Saccharomyces cerevisiae) and consisting of protein-carbohydrate complexes is frequently used as a pathogen in phagocytosis assays.

Application

The Phagocytosis Assay Kit (Green Zymosan) uses fluorescently pre-labeled Zymosan particles to detect and quantify in vitro phagocytosis by fluorescent microscope, spectrophotometer (Ex/Em = 490/520 nm) or flow cytometry.

Suitability

This kit is suitable for screening activators and/or inhibitors of phagocytosis and Toll-like receptor ligands (TLR) in mammalian cells.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service