CLS480116
Corning® LSE™ single block, for Corning LSE single block digital dry baths
single block, 48 x 0.2 mL PCR tubes or 6 x 0.2 mL strips, 1/cs
Synonym(s):
equipment replacement accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NC.21
Recommended Products
feature
Holds 48 x 0.2 mL tubes
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 480116
technique(s)
PCR: suitable
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
CLS6785SB, CLS6786SB, CLS6787SB
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service