CLS45006L
Corning® ProCulture® glass spinner flask complete, baffled, angled sidearms
6 L glass spinner flask, w/ 100 mm flat center cap, 2 45 mm vertical sidearms, reusable, non-sterile, 1/cs
Synonym(s):
cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
About This Item
Recommended Products
material
borosilicate glass (borosilicate glass code 7740)
polypropylene cap
screw top cap
three-neck
sterility
non-sterile
feature
cap
closure type Screw cap
reusable
flask style: Spinner
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 4500-6L
capacity
6 L
neck diam.
100 mm
side arm diam.
45 mm
working volume
6 L
6000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® glass flask and other parts constructed of FDA-approved materials
- baffles break up laminar flow and improve circulation
- unique paddle design eliminates dead spots, agitates surface for better aeration
- wide center neck forms visible airtight seal
- two angled sidearms offer easy access without interrupting culture
- excellent growth of cells in suspension as well as anchorage-dependent cells growing on microcarrier beads
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service