Key Documents
BMSIPS2050PT1
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
flat blade, PTFE-coated
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
Pricing and availability is not currently available.
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-T1
diam. × L
60 mm × 400 mm
General description
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
This one line, PTFE coated propeller is useful for low viscosity, mid or high speed mixing.
Application
stirring, mixing
Choose from one of the most recent versions:
Certificates of Analysis (COA)