Skip to Content
Merck
All Photos(2)

Documents

AV37905

Sigma-Aldrich

Anti-MyBBP1A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AL024407, Anti-AU019902, Anti-MyB binding protein (P160) 1a, Anti-P160, Anti-p160MBP, Anti-p67MBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

152 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Mybbp1a(18432)

General description

MyB binding protein (P160) 1a (MyBBP1A), a c-myb proto-oncogene product (c-Myb)-interacting protein, is post-translationally processed to 67 kDa fragment (p67MBP). MyBBP1A is primarily expressed in the nucleoli. MyBBP1A which is associated with RNA Polymerase 1 complex and ribosome biogenesis machinery regulates rRNA metabolism and is believed to connect the process of ribosome biogenesis and Myb-dependent transcription to regulate/coordinate cell cycle progression and proliferation.

Specificity

Anti-MyBBP1A polyclonal antibody reacts with human, rat, and mouse MyB binding protein (P160) 1a proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of mouse Mybbp1a

Application

Anti-MyBBP1A polyclonal antibody is used to tag MyB binding protein (P160) 1a protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of MyB binding protein (P160) 1a in the coordination and regulation of rRNA biosynthesis and ribosome biogenesis.

Biochem/physiol Actions

Mybbp1a may activate or repress transcription via interactions with sequence specific DNA-binding proteins. Repression may be mediated at least in part by histone deacetylase activity.

Sequence

Synthetic peptide located within the following region: HSSGSNRLYDLYWQAMRMLGVQRPKSEKKNAKDIPSDTQSPVSTKRKKKG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service