Skip to Content
Merck
All Photos(1)

Key Documents

AV35764

Sigma-Aldrich

Anti-DLX6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Distal-less homeobox 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

dog, bovine, mouse, horse, rabbit, rat, human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLX6(1750)

Immunogen

Synthetic peptide directed towards the middle region of human DLX6

Biochem/physiol Actions

Distal-less homeobox 6 (DLX6) is a transcription factor that regulates craniofacial, forebrain and limb and genital bud development. DLX proteins have multiple functions at different stages of development and are expressed in differentiating osteoblasts and placental formation.

Sequence

Synthetic peptide located within the following region: ENGEIRFNGKGKKIRKPRTIYSSLQLQALNHRFQQTQYLALPERAELAAS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond F Robledo et al.
Genes & development, 16(9), 1089-1101 (2002-05-10)
Dlx homeobox genes are mammalian homologs of the Drosophila Distal-less (Dll) gene. The Dlx/Dll gene family is of ancient origin and appears to play a role in appendage development in essentially all species in which it has been identified. In
G R Merlo et al.
The International journal of developmental biology, 44(6), 619-626 (2000-11-04)
Dlx genes comprise a highly conserved family of homeobox genes homologous to the distal-less (Dll) gene of Drosophila. They are thought to act as transcription factors. All Dlx genes are expressed in spatially and temporally restricted patterns in craniofacial primordia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service