Skip to Content
Merck
All Photos(1)

Key Documents

A111060

Sigma-Aldrich

DMT-dA(bz) Phosphoramidite

configured for ABI 39X

Synonym(s):

DMT-dA(bz) Amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyadenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H52N7O7P
CAS Number:
Molecular Weight:
857.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

description

color-coded label

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)

color

white to off-white

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI 39X

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C47H52N7O7P/c1-32(2)54(33(3)4)62(59-27-13-26-48)61-40-28-42(53-31-51-43-44(49-30-50-45(43)53)52-46(55)34-14-9-7-10-15-34)60-41(40)29-58-47(35-16-11-8-12-17-35,36-18-22-38(56-5)23-19-36)37-20-24-39(57-6)25-21-37/h7-12,14-25,30-33,40-42H,13,27-29H2,1-6H3,(H,49,50,52,55)/t40-,41+,42+,62?/m0/s1

InChI key

GGDNKEQZFSTIMJ-AKWFTNRHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dA(bz) Phosphoramidite belongs to the DNA Phosphoramidites group.The key features of standard protected DNA phosphoramidite are:
  • Exocyclic amine functions are protected by a benzoyl group (dA(bz) and dC(bz)) or isobutyryl group (dG(ib))
  • Recommended cleavage and deprotection conditions are 8 hours at 55 °C or 24 hours at room temperature using concentrated ammonia solution for standard base-protected oligonucleotides
  • The high coupling efficiency of Proligo′s DNA phosphoramidites leads to high-yield and high-quality oligonucleotides

Packaging

packaged in a 60 ml septum bottle

Legal Information

ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service