Skip to Content
Merck
All Photos(1)

Documents

15167

Sigma-Aldrich

NIR-797 isothiocyanate

suitable for fluorescence, ≥70% (coupling to amines)

Synonym(s):

1,1′-Bis(4-sulfobutyl)-11-(4-isothiocyanatophenylthio)-3,3,3′,3′-tetramethyl-10,12-tri methyleneindotricarbocyanine monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H50N3NaO6S4
CAS Number:
Molecular Weight:
880.14
Beilstein:
6681910
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥70% (coupling to amines)

form

powder

fluorescence

λex 795 nm; λem 817 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Na+].CC1(C)/C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCS([O-])(=O)=O)c4ccccc4C3(C)C)=C2Sc5ccc(cc5)N=C=S)N(CCCCS([O-])(=O)=O)c6ccccc16

InChI

1S/C45H51N3O6S4.Na/c1-44(2)37-16-5-7-18-39(37)47(28-9-11-30-57(49,50)51)41(44)26-20-33-14-13-15-34(43(33)56-36-24-22-35(23-25-36)46-32-55)21-27-42-45(3,4)38-17-6-8-19-40(38)48(42)29-10-12-31-58(52,53)54;/h5-8,16-27H,9-15,28-31H2,1-4H3,(H-,49,50,51,52,53,54);/q;+1/p-1

InChI key

URQBXUCJHTXMKS-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Near-infrared cyanine dyes for labeling of proteins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Lipowska et al.
Synthetic Communications, 23, 3087-3087 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service