Skip to Content
Merck
All Photos(1)

Key Documents

151-500

Sigma-Aldrich

Skeletal Muscle Cell Growth Medium (500ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Skeletal Muscle Cell Growth Medium (500ml) has been used to culture undifferentiated human skeletal muscle cells.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Jean Wang et al.
Life sciences, 82(1-2), 11-20 (2007-12-11)
Riluzole is known to be of therapeutic use in the management of amyotrophic lateral sclerosis. In this study, we investigated the effects of riluzole on ion currents in cultured differentiated human skeletal muscle cells (dHSkMCs). Western blotting revealed the protein
Sirada Srihirun et al.
Nitric oxide : biology and chemistry, 94, 1-8 (2019-10-12)
Several studies show that dietary nitrate enhances exercise performance, presumably by increasing muscle blood flow and improving oxygen utilization. These effects are likely mediated by nitrate metabolites, including nitrite and nitric oxide (NO). However, the mechanisms of nitrate production, storage

Protocols

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service