Skip to Content
Merck
All Photos(1)

Key Documents

60152

Sigma-Aldrich

Chromium(III) potassium sulfate dodecahydrate

purum p.a., ≥98.5% (RT)

Synonym(s):

Chrome alum, Potassium chromium(III) sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
499.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥98.5% (RT)

form

crystals

mp

89 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤200 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Cr.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

ZFVHBEKVAITXHW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chromium(III) potassium sulfate dodecahydrate (Potassium chromium(III) sulfate dodecahydrate, Chrome alum) is a violet colored solid. It has been synthesized by reacting sulfur dioxide with potassium dichromate (diisolved in sulfuric acid) at -38°C. It is widely used chemical in dyeing and textile industry.

Application

Chromium(III) potassium sulfate dodecahydrate (KCr(SO4)2.12H2O) may be used for coating the slides for microscopic studies on Periplaneta americana. It may be used in the preparation of quasi-monodisperse spherical core-shell particles of Cr/α-Cr2O3.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Black Cr/α-Cr2O3 nanoparticles based solar absorbers.
Khamlich S, et al.
Physica B: Condensed Matter, 407(10), 1509-1512 (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 887-887 (1994)
G M Lee et al.
The Histochemical journal, 17(11), 1221-1233 (1985-11-01)
Various pretreatments of metaphase spreads were examined to obtain optimal DNA labelling patterns while maintaining chromosome integrity during in situ hybridization procedures. Preparations of African green monkey (AGM) chromosomes fixed in methanol-acetic acid (CV-1 cell line) were treated by coating
V M Chikov et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 294-297 (1989-03-01)
Gallocyanine-chrome alum-stained pig embryonic kidney cells have paramagnetic properties. They move under the influence of gradient magnetic field (magnetophoresis). The velocity of magnetophoresis is proportional to the content of nucleic acids in cells. This allows to estimate the content of
O Schmitt et al.
Micron (Oxford, England : 1993), 28(3), 197-215 (1997-06-01)
The investigation of neurohistological specimens by image analysis has become an important tool in morphological neuroscience. The problems which arise during the processing of these images are non-trivial, especially if a pattern recognition of cells in the imaged tissue is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service