Skip to Content
Merck
All Photos(4)

Key Documents

86195

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

purum p.a., ≥98.0% (T)

Synonym(s):

2-Hydroxy-5-sulfobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein:
650741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥98.0% (T)

form

solid

mp

105-110 °C (lit.)

solubility

water: soluble 127.1 g/L at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O

InChI

1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2

InChI key

BHDKTFQBRFWJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Sulfosalicylic acid is strongly acidic in nature. It exists in various hydrated forms such as dihydrate, dideuterate, trihydrate and pentahydrate. It acts as a ligand and forms various coordination complexes. It forms a complex with theophylline. Crystal structure of the complex was investigated by X-ray photoelectron spectroscopy (XPS).

Application

5-Sulfosalicylic acid dihydrate may be used in the preparation of 1:1 proton-transfer organic adduct, 3-aminopyridin­ium 3-carb­oxy-4-hydroxy­benzene­sulfonate monohydrate and the anhydrous adduct.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two salts of 5-sulfosalicylic acid and 3-aminopyridine.
Meng XG, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 63(11), o667-o670 (2007)
Poly [[tris (di-2-pyridylamine) tris (μ-5-sulfonatosalicylato) tricopper (II)] trihydrate].
Fan SR and Zhu LG.
Acta Crystallographica Section E, Structure Reports Online, 61(1), m174-m176 (2004)
Hydrogen bonding in proton-transfer compounds of 5-sulfosalicylic acid with ortho-substituted monocyclic heteroaromatic Lewis bases.
Smith G, et al.
Journal of Chemical Crystallography, 36(12), 841-849 (2006)
Joanna S Stevens et al.
The journal of physical chemistry. B, 114(44), 13961-13969 (2010-10-22)
Recent studies suggested that X-ray photoelectron spectroscopy (XPS) sensitively determines the protonation state of nitrogen functional groups in the solid state, providing a means for distinguishing between co-crystals and salts of organic compounds. Here we describe how a new theophylline
M Vázquez et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 74, 349-359 (2014-12-03)
The mercurial forms [inorganic divalent mercury, Hg(II) and methylmercury, CH3Hg] produce neurological and immune effects as well as hematological and renal alterations. The main route of exposure is through the diet. Consequently, the gastrointestinal mucosa is exposed to these mercurial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service