Skip to Content
Merck
All Photos(1)

Key Documents

69530

Supelco

2-Ethylfuran

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.437-1.443
n20/D 1.439 (lit.)

bp

92-93 °C/768 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCc1ccco1

InChI

1S/C6H8O/c1-2-6-4-3-5-7-6/h3-5H,2H2,1H3

InChI key

HLPIHRDZBHXTFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco J Hidalgo et al.
Journal of agricultural and food chemistry, 62(49), 12045-12051 (2014-11-25)
The carbonyl-scavenging ability of 2-amino-1-methylbenzimidazole (AMBI) and the heterocyclic aromatic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) was investigated in an attempt to identify new routes that can modify the carbonyl content of foods. The reaction of both AMBI and PhIP with 2-alkenals, 2,4-alkadienals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service