Skip to Content
Merck
All Photos(1)

Key Documents

49129

Sigma-Aldrich

Carbon disulfide

puriss., low in benzene, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

vapor density

2.67 (vs air)

Quality Level

vapor pressure

5.83 psi

grade

puriss.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

212 °F

quality

low in benzene

expl. lim.

50 %

impurities

≤1 ppm benzene
≤1 ppm toluene

refractive index

n20/D 1.627 (lit.)
n20/D 1.627

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.263 g/mL at 20 °C
1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marjan J Smeulders et al.
Nature, 478(7369), 412-416 (2011-10-21)
Extremophilic organisms require specialized enzymes for their exotic metabolisms. Acid-loving thermophilic Archaea that live in the mudpots of volcanic solfataras obtain their energy from reduced sulphur compounds such as hydrogen sulphide (H(2)S) and carbon disulphide (CS(2)). The oxidation of these
[11C]carbon disulfide: a versatile reagent for PET radiolabelling.
Philip W Miller et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(2), 433-436 (2011-12-14)
Margherita Maiuri et al.
Physical chemistry chemical physics : PCCP, 14(18), 6312-6319 (2012-02-15)
In carotenoids internal conversion between the allowed (S(2)) and forbidden (S(1)) excited states occurs on a sub-picosecond timescale; the involvement of an intermediate excited state(s) (S(x)) mediating the process is controversial. Here we use high time resolution (sub-20 fs) broadband
Mark B van Eldijk et al.
Chemical communications (Cambridge, England), 49(71), 7770-7772 (2013-06-19)
CS2 hydrolase, a zinc-dependent enzyme that converts carbon disulfide to carbon dioxide and hydrogen sulfide, exists as a mixture of octameric ring and hexadecameric catenane forms in solution. A combination of size exclusion chromatography, multi-angle laser light scattering, and mass
Continuous direct spinning of fibers of single-walled carbon nanotubes with metallic chirality.
Rajyashree M Sundaram et al.
Advanced materials (Deerfield Beach, Fla.), 23(43), 5064-5068 (2011-10-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service