Skip to Content
Merck
All Photos(1)

Key Documents

32915

Supelco

Fumonisin B2-13C34 solution

~10 μg/mL in acetonitrile: water, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C34H59NO14
CAS Number:
Molecular Weight:
739.58
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~10 μg/mL in acetonitrile: water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+34

storage temp.

−20°C

SMILES string

[13CH3][13CH2][13CH2][13CH2][13C@@H]([13CH3])[13C@@H](O[13C](=O)[13CH2][13C@@H]([13CH2][13C](O)=O)[13C](O)=O)[13C@H]([13CH2][13C@@H]([13CH3])[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C@@H](O)[13CH2][13C@H](O)[13C@H]([13CH3])N)O[13C](=O)[13CH2][13C@@H]([13CH2][13C](O)=O)[13C](O)=O

InChI key

UXDPXZQHTDAXOZ-BMUNAYDHSA-N

General description

This is the 13C isotope-labelled analog of mycotoxin, Fumonisin B2. Both labelled and non-labelled single-component solutions are offered for precise quality control of food and feed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service