VSWP04700
MCE Membrane Filter, 0.025 μm Pore Size
MF-Millipore™, filter diam. 47 mm, hydrophilic
Synonym(s):
Hydrophilic mixed cellulose esters (MCE) membrane filter discs, MF-Millipore™ Membrane Filter, 0.025 µm pore size
About This Item
Recommended Products
material
mixed cellulose esters (MCE) membrane
plain filter
white filter
Quality Level
description
47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
MF-Millipore™
Millipore
parameter
0.15 L/min-cm2 air flow rate
0.15 mL/min-cm2 water flow rate
75 °C max. temp.
filter diam.
47 mm
thickness
100 μm
gravimetric extractables
1.5%
refractive index
n/D 1.5
matrix
MF-Millipore™
pore size
0.025 μm pore size
72 % porosity
bubble point
≥21.1 bar, air with water at 23 °C
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: General laboratory filtration
- Intended Use: Retention or removal of biological contaminants
- Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
- Storage Statement: Store in dry location away from heat source
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
This article is a protocol for Simple dialysis of small volumes using V Series membranes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service