SPR0LS001
SmartPak® Purification Pack
For use with RiOs™ 3/5 Water Purification System
Synonym(s):
SmartPak® RO 5 Purification Pack LT
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
SmartPak®
compatibility
for use with RiOs™
Looking for similar products? Visit Product Comparison Guide
General description
This replacement cartridge is used to pretreat water by efficiently removing contaminants from feed water.
Application
The single SmartPak® RO cartridge (with integrated RO membrane) enables easy and rapid replacement.
Legal Information
RIOS is a trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service