MHA00FCR2
m-FC Broth without Rosolic Acid
Ampoule used for testing fecal coliforms in waste and effluent water
Synonym(s):
m-FC Broth
About This Item
Quality Level
sterility
sterile
form
liquid
shelf life
12 mo.
feature
ready-to-use
packaging
pack of 50 tests
capacity
2 mL
filling volume
2 mL
color
slightly opalescent blue
input
sample type bottled water
sample type wine
sample type cosmetics
sample type sport drink
sample type soft drink
sample type food(s)
sample type beverage(s)
sample type cider
sample type beer
pH
7.4±0.2 (25.0 °C)
application(s)
environmental
industrial qc
microbiology
water monitoring
compatibility
for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®
shipped in
wet ice
storage temp.
2-8°C
suitability
fecal coliforms
General description
Application
Packaging
Specifications
Culture Medium
Analysis Note
E. coli (ATCC 25922)
Mixed culture (E. coli, P. aeruginosa and P. vulgaris)
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service