Skip to Content
Merck
All Photos(1)

Key Documents

MAB4145

Sigma-Aldrich

Anti-BCRP Antibody, clone BXP-34

culture supernatant, clone BXP-34, Chemicon®

Synonym(s):

Breast Cancer Resistance Protein, Placenta-specific ATP Binding Cassette Transporter, ATP-binding Cassette, Sub Family G, member 2 (ABCG2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

BXP-34, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)

isotype

IgG1

suitability

not suitable for Western blot
not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCG2(9429)

General description

The 72 kDa breast cancer resistance protein (BCRP) is the second member of the subfamily G of the human ATP binding cassette (ABC) transporter superfamily and thus also designated as ABCG2. BCRP is a half-transporter consisting of only 1 nucleotide binding domain followed by 1 membrane-spanning domain. Current experimental evidence suggests that BCRP may function as a homodimer or homotetramer. Overexpression of BCRP is associated with high levels of resistance to a variety of anticancer agents. BCRP expression has been detected in a large number of hematological malignancies and solid tumors, indicating that this transporter may play an important role in clinical drug resistance of cancers. In addition to its role to confer resistance against chemotherapeutic agents, BCRP actively transports structurally diverse organic molecules, conjugated or unconjugated, such as estrone-3-sulfate, 17-β-estradiol 17-(β-D-glucuronide), and methotrexate. BCRP is highly expressed in the placental syncytiotrophoblasts, in the apical membrane of the epithelium in the small intestine, in the liver canalicular membrane, and at the luminal surface of the endothelial cells of human brain microvessels. This strategic and substantial tissue localization indicates that BCRP also plays an important role in absorption, distribution, and elimination of drugs that are BCRP substrates.

Specificity

This antibody reacts with an internal epitope of the Breast Cancer Resistance Protein (BCRP), a 70 kDa transmembrane half-transporter which is involved in multidrug resistance. The antibody does not cross-react with human MDR1, MRP1, MRP2 or MRP5 gene products.

Immunogen

The Mitoxanthrone resistant, BCRP-overexpressing cell line MCF7-MR.

Application

Detect BCRP using this Anti-BCRP Antibody, clone BXP-34 validated for use in IC, IH.
Immunocytochemistry: 1:20 - 1:50 on acetone fixed cytospin preparations

Immunohistochemistry: 1:20 on acetone fixed frozen tissue sections

Not suitable for paraffin-embedded tissues or for Western blotting

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Target description

70 kDa

Physical form

Liquid containing 0.7% BSA and 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for up to 6 months. For long-term storage store at -20°C.

Analysis Note

Control
MCF-7 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Paprocka et al.
European journal of haematology, 99(5), 415-422 (2017-08-15)
It is well established that expression of multi-drug resistance (MDR) proteins (MDR1, BCRP, MDR3, MRP1, and LRP) in leukemic blasts correlates with acute myeloid leukemia (AML) patients' clinical response. Assuming that leukemic stem cells (LSC) are resistant to chemotherapy and
Wen Jing et al.
Oncology letters, 20(6), 309-309 (2020-10-24)
Most patients with advanced leukemia eventually die from multidrug resistance (MDR). Chemotherapy-resistant leukemia cells may lead to treatment failure and disease relapse. Overexpression of ATP-binding cassette subfamily G member 2 (ABCG2) leads to MDR, which serves as a potential biomarker

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service