Skip to Content
Merck
All Photos(1)

Documents

MAB1953Z

Sigma-Aldrich

Anti-Integrin αV Antibody, clone P3G8, azide free

clone P3G8, Chemicon®, from mouse

Synonym(s):

CD51, MAB1953

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P3G8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Specificity

Reacts with human alphaV integrin subunit and with all alphaV-containing integrin receptors.

Immunogen

UCLA P3 lung carcinoma cells

Application

Immunohistochemistry. For use on acetone or paraformaldehyde fixed tissue

Immunocytochemistry. Will react with some lymphoid cell lines (B cells), many carcinoma and melanoma cell lines and osteosarcomas.

Flow cytometry

Immunoprecipitation

EIA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin αV Antibody, clone P3G8, azide free is validated for use in ELISA, FC, IP, IC, IH for the detection of Integrin αV.

Physical form

Format: Purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony H T Jeremy et al.
The Journal of investigative dermatology, 121(1), 20-27 (2003-07-04)
The earliest subclinical acne "lesion" is a microcomedone, of which hyperproliferation of the follicular epithelium is a characteristic feature. Inflammatory cells have been observed at the periphery of these "lesions". This study investigated whether inflammatory events occur pre or post
Paul A Rupp et al.
Development (Cambridge, England), 131(12), 2887-2897 (2004-05-21)
A major challenge confronting developmental cell biologists is to understand how individual cell behaviors lead to global tissue organization. Taking advantage of an endothelial cell-specific marker and scanning time-lapse microscopy, we have examined the formation of the primary vascular pattern
Divya Varun et al.
Acta biomaterialia, 48, 120-130 (2016-12-19)
Despite therapeutic advances, neurodegenerative diseases and disorders remain some of the leading causes of mortality and morbidity in the United States. Therefore, cell-based therapies to replace lost or damaged neurons and supporting cells of the central nervous system (CNS) are
Chun-Chieh Hsu et al.
Scientific reports, 6, 23387-23387 (2016-03-19)
Bacterial infection-induced sepsis is the leading cause of septic inflammatory disease. Rhodostomin (Rn), a snake venom disintegrin, was previously reported to interact with the αVβ3 integrin and the TLR4 on phagocyte in attenuating LPS-induced endotoxemia. In this report, we further
Vladimir Leksa et al.
The Journal of biological chemistry, 287(27), 22450-22462 (2012-05-23)
The plasminogen (Plg) activation cascade on the cell surface plays a central role in cell migration and is involved in a plethora of physiological and pathological processes. Its regulation is coordinated by many receptors, in particular the urokinase-type plasminogen activator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service