Skip to Content
Merck
All Photos(1)

Key Documents

8.21092

Sigma-Aldrich

Tetrahydrofurfuryl alcohol polyethylene glycol ether

for synthesis

Synonym(s):

Tetrahydrofurfuryl alcohol polyethylene glycol ether, Glycofurol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.22

Quality Level

form

liquid

bp

176 °C/40 hPa

density

1.08 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(CCC1)COCCO

InChI

1S/C7H14O3/c8-3-5-9-6-7-2-1-4-10-7/h7-8H,1-6H2

InChI key

CTPDSKVQLSDPLC-UHFFFAOYSA-N

Application


  • Biodegradable microparticle production: A novel green process employing CO2 medium was developed for the production of biodegradable microparticles encapsulating TGF-beta1, utilizing tetrahydrofurfuryl alcohol polyethylene glycol ether, enhancing the sustained release properties of the therapeutic proteins (Swed et al., 2015).

  • Nanocarrier devices for targeted drug delivery: Tetrahydrofurfuryl alcohol polyethylene glycol ether was utilized in the formulation of photosensitizer-loaded nanocarrier devices, bridging laboratory and large-scale production, and demonstrating potential in targeted drug delivery systems (Beyer et al., 2015).

  • Cardiac tissue-engineering: In cardiac tissue engineering, tetrahydrofurfuryl alcohol polyethylene glycol ether was incorporated into a thermosensitive hydrogel for releasing growth factors from pharmacologically active microcarriers, presenting a biomimetic 3D scaffold that supports cardiac cell growth (Karam et al., 2014).

  • Enhanced pharmacokinetics in drug delivery: A self-microemulsifying drug delivery system incorporating tetrahydrofurfuryl alcohol polyethylene glycol ether demonstrated improved pharmacokinetics for tacrolimus, suggesting its efficacy in bioavailability enhancement (von Suesskind-Schwendi et al., 2013).

Analysis Note

Density (d 20 °C/ 4 °C): 1.080 - 1.090
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service