Skip to Content
Merck
All Photos(1)

Key Documents

8.01962

Sigma-Aldrich

Brij® 35

for synthesis

Synonym(s):

Brij® 35, Polyoxyethylene lauryl ether, Polyethyleneglycol lauryl ether

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
(C2H4O)nC12H26O
CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
500-002-6
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

solid

potency

>78000 mg/kg LD50, oral (Rat)

pH

5.5-7.0 (20 °C, 100 g/L in H2O)

bp

100 °C/1013 hPa

transition temp

flash point >149 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCO)CCCCCCCCCCCC

Looking for similar products? Visit Product Comparison Guide

Application

  • Application of artificial neural networks for enhancing Aspergillus flavipes lipase synthesis for green biodiesel production.: This study explores the use of artificial neural networks to enhance the synthesis of lipase by Aspergillus flavipes, which is crucial for green biodiesel production. The use of Brij® 35 in the optimization process highlights its effectiveness as a nonionic surfactant in biochemical applications (El-Metwally et al., 2023).
  • Solvents and detergents compatible with enzyme kinetic studies of cholinesterases.: This research investigates various solvents and detergents, including Brij® 35, for their compatibility in enzyme kinetic studies involving cholinesterases, underlining the utility of Brij® 35 in biochemical and pharmacological research (Sands et al., 2023).
  • Development of green micellar HPLC-DAD method for simultaneous determination of some sulbactam combinations used in COVID-19 regimen.: The study develops a green micellar high-performance liquid chromatography method using Brij® 35 for the analysis of sulbactam combinations, showcasing its role in pharmaceutical formulations and chemical analysis (Ragab et al., 2023).
  • Enhanced fluorimetric detection of diphenylpyraline HCl using micelle and cyclodextrin mediated approach: Spectrofluorimetric and micellar liquid chromatographic application for either single or combined formulation with caffeine and paracetamol.: This paper discusses the enhancement of fluorimetric detection of diphenylpyraline HCl through the use of Brij® 35, demonstrating its effectiveness in improving analytical sensitivity in pharmaceutical applications (Hammad et al., 2023).
  • Application of micellar liquid chromatography to model ecotoxicity of pesticides. Comparison with immobilized artificial membrane chromatography and n-octanol-water partitioning.: This research uses Brij® 35 in micellar liquid chromatography to model the ecotoxicity of pesticides, emphasizing its applicability in environmental chemistry and toxicology studies (Stergiopoulos et al., 2023).

Analysis Note

Hydroxyl value: 45 - 50
Water (K. F.): ≤ 3.0 %
Identity (IR): passes test

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

300.2 °F

Flash Point(C)

> 149 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service