8.00836
Ethyl acrylate
(stabilised with hydroquinone monomethyl ether) for synthesis
Synonym(s):
Ethyl acrylate, Acrylic acid ethyl ester
About This Item
synthesis grade
Recommended Products
grade
for synthesis
synthesis grade
Quality Level
vapor pressure
39 hPa ( 20 °C)
Assay
≥99% (GC)
form
liquid
autoignition temp.
355 °C (DIN 51794)
potency
550 mg/kg LD50, oral (Rat)
1800 mg/kg LD50, skin (Rabbit)
expl. lim.
1.8-14 % (v/v)
dilution
(for synthesis)
kinematic viscosity
0.582 cSt(25 °C)
bp
100 °C/1013 hPa
mp
-71 °C
transition temp
flash point 9 °C
solubility
20 g/L
density
0.92 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CC)C(=O)C=C
InChI
1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3
InChI key
JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.921 - 0.922
Identity (IR): passes test
The material is stabilised with MEHQ. At time of analytical testing a content of >10 ppm stabiliser was determined.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
48.2 °F - closed cup
Flash Point(C)
9 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service