Skip to Content
Merck
All Photos(1)

Key Documents

8.00097

Sigma-Aldrich

Dimethyl adipate

for synthesis

Synonym(s):

Dimethyl adipate, Dimethyl hexanedioate, Adipic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OOC(CH2)4COOCH3
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-020-6
NACRES:
NA.22

vapor pressure

0.06 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

360 °C

potency

8191 mg/kg LD50, oral (Rat)
>2250 mg/kg LD50, skin (Rabbit)

expl. lim.

0.81-8.1 % (v/v)

kinematic viscosity

2.5 cSt(25 °C)

bp

215-225 °C/1013 hPa

mp

8-11 °C

transition temp

flash point 107 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3

InChI key

UDSFAEKRVUSQDD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.060 - 1.063
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service