Skip to Content
Merck
All Photos(1)

Key Documents

345743

Sigma-Aldrich

Disialoganglioside-GD₂ from bovine brain

≥98% (TLC), lyophilized, cell type and antigen marker, Calbiochem®

Synonym(s):

Ganglioside GD2, Disialo, Human Brain, GalNAcβ1,4(Neu5Acα2,8)(Neu5Acα2,03)Galβ1,4Glcβ1,1ʹ Ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H138N4O34
CAS Number:
Molecular Weight:
1675.94
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

product name

Ganglioside GD2, Disialo, Human Brain, Ganglioside GD₂, Disialo, Human Brain, CAS 65988-71-8, is a highly purified sialic acid-containing glycolipids that is useful as markers of various cell types and antigens.

Quality Level

Assay

≥98% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform/methanol (2:1): soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C74H134N4O32/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-52(89)78-43(44(84)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)40-101-69-61(95)60(94)63(50(38-81)104-69)106-70-62(96)67(64(51(39-82)105-70)107-68-55(77-42(3)83)59(93)58(92)49(37-80)103-68)110-74(72(99)100)35-46(86)54(76)66(109-74)57(91)48(88)41-102-73(71(97)98)34-45(85)53(75)65(108-73)56(90)47(87)36-79/h30,32,43-51,53-70,79-82,84-88,90-96H,4-29,31,33-41,75-76H2,1-3H3,(H,77,83)(H,78,89)(H,97,98)(H,99,100)/b32-30+/t43?,44?,45-,46-,47?,48?,49-,50-,51-,53+,54+,55-,56?,57?,58+,59-,60-,61-,62-,63-,64+,65+,66+,67-,68+,69-,70+,73-,74+/m1/s1

InChI key

FFILOTSTFMXQJC-QCFYAKGBSA-N

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful as markers of various cell types and antigens
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumeyye Cavdarli et al.
Cells, 10(6) (2021-07-03)
The O-acetylated form of GD2, almost exclusively expressed in cancerous tissues, is considered to be a promising therapeutic target for neuroectoderm-derived tumors, especially for breast cancer. Our recent data have shown that 9-O-acetylated GD2 (9-OAcGD2) is the major O-acetylated ganglioside

Articles

Glycosphingolipids

Glycosphingolipids

Glycosphingolipids

Glycosphingolipids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service