Skip to Content
Merck
All Photos(1)

Key Documents

1.09887

Supelco

Buffer concentrate

(phosphate), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 7.00 ± 0.02 (20°C) Titrisol®

Synonym(s):

Buffer concentrate for 500 ml buffer solution, pH 7.00 ± 0.02 (20°C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161706
NACRES:
NA.25

grade

analytical standard

Quality Level

Agency

according to DIN 19266

product line

Titrisol®

pH

7.00 (20 °C in H2O)

density

1.08 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The Titrisol® reference buffer concentrate [phosphate; pH=7.00±0.02 (20 °C)] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266. The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.

Application


  • A novel and efficient voltammetric sensor for the simultaneous determination of alizarin red S and tartrazine: Utilizing buffer concentrates, this study develops a poly(leucine) functionalized carbon paste electrode. It demonstrates the crucial role of buffer solutions in enhancing the accuracy and sensitivity of voltammetric sensors in environmental and biochemical assays (Moulya et al., 2024).

  • Nitrocellulose for Prolonged Permeation of Levofloxacin HCl-Salicylic Acid In Situ Gel: Research grade buffer concentrate is employed to maintain pH stabilization during the formulation of nitrocellulose-based in situ gels, improving drug delivery systems for pharmaceutical applications (Khaing et al., 2024).

  • The development of lateral flow devices for urinary biomarkers to assess kidney health: This study integrates high-purity buffer concentrates for the optimal performance of lateral flow devices, underscoring their importance in clinical diagnostics and biochemical assays (D′Souza et al., 2024).

  • Determination of lamotrigine in human plasma by HPLC-PDA: Buffer concentrates are crucial in the forensic analysis of lamotrigine levels in plasma, illustrating their significance in enhancing the reliability and accuracy of pharmaceutical testing (Sánchez-Sellero et al., 2024).

  • On-Chip Electrochemical Sensing with an Enhanced Detecting Signal Due to Concentration Polarization-Based Analyte Preconcentration: The use of buffer concentrates is essential in creating on-chip electrochemical sensors, facilitating significant advancements in analytical chemistry through improved detection signals (Park et al., 2024).

Features and Benefits

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive and batch-specific Certificate of Analysis.

Analysis Note

pH-value (20 °C): 7.00 +/- 0.02
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

standard

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service