Skip to Content
Merck
All Photos(1)

Key Documents

1.06174

Millipore

TAE buffer

50 x pH 8,3 tris-acetate EDTA buffer

Synonym(s):

50X TAE running buffer, Tris-Acetate EDTA buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

form

liquid

pH

8.2-8.4 (20 °C in H2O)

density

1.1 g/cm3 at 20 °C

storage temp.

no temp limit

Application


  • Detection of Dengue Virus From Aedes aegypti (Diptera, Culicidae) in Field-Caught Samples From Makkah Al-Mokarramah, Kingdom of Saudi Arabia, Using RT-PCR.: The use of TAE buffer in RT-PCR protocols for detecting Dengue virus in field samples underscores its importance in virological studies and public health research (Ali et al., 2022).

  • Semi-Automatic Lab-on-PCB System for Agarose Gel Preparation and Electrophoresis for Biomedical Applications.: This paper describes a lab-on-PCB system that automates agarose gel preparation and electrophoresis, employing TAE buffer, thereby streamlining these processes in biomedical research (Urbano-Gámez et al., 2021).

  • A facile thermometer-like electrophoresis titration biosensor for alternative miRNA assay via moving reaction boundary chip.: This study presents a novel biosensor for miRNA assays, utilizing TAE buffer in electrophoresis, enhancing the precision and efficiency of molecular diagnostics (Chen et al., 2021).

Analysis Note

Appearance: clear and colourless
pH-value: 8.2 - 8.4

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service