Skip to Content
Merck
All Photos(1)

Documents

1.05680

Supelco

Lithium carbonate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Lithium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CLi2O3
CAS Number:
Molecular Weight:
73.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
209-062-5
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

525 mg/kg LD50, oral (Rat)

impurities

≤0.01% Substances insoluble in dilute hydrochloric acid

pH

10-11 (20 °C, 5 g/L in H2O)

mp

720 °C

solubility

13 g/L

density

2.1 g/cm3 at 20 °C

bulk density

250 kg/m3

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.0005%
sulfur compounds (as SO42-): ≤0.2%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Application


  • Radical Polymer-based Positive Electrodes for Dual-Ion Batteries: Describes the enhancement of battery performance through the use of γ-Butyrolactone-based electrolytes in conjunction with radical polymer-based positive electrodes, highlighting advances in lithium carbonate applications in battery technology (Rudolf K, et al., 2024).

  • Analysis of Trace Impurities in Lithium Carbonate: Focuses on the precision analysis of impurities within pharmaceutical grade lithium carbonate, crucial for maintaining high purity standards in pharmaceutical applications (Suárez A, et al., 2024).

  • Effects of Central Metal Ion on Binuclear Metal Phthalocyanine-Based Redox Mediator for Lithium Carbonate Decomposition: Investigates the role of central metal ions in enhancing the efficiency of lithium carbonate decomposition, important for environmental lithium analysis and recovery (Yan Q, et al., 2024).

  • Selective preparation of lithium carbonate from overhaul slag by high temperature sulfuric acid roasting - Water leaching: Details a method for extracting high-purity lithium carbonate from industrial waste, demonstrating an environmentally friendly approach to lithium recovery (Dong L, et al., 2024).

  • Reactive molecular dynamics simulations of lithium-ion battery electrolyte degradation: Explores the degradation mechanisms of lithium-ion battery electrolytes using simulations, contributing to the development of more durable lithium battery materials (Mabrouk Y, et al., 2024).

Linkage

Replaces: LX0325-1

Analysis Note

  • Assay(acidimetric): ≥ 99.0 %
  • Substances insoluble in dilute hydrochloric acid: ≤ 0.01 %
  • Chloride (Cl): ≤ 0.002 %
  • Nitrate (NO₃): ≤ 0.0005 %
  • Sulfur compounds (as SO₄): ≤ 0.2 %
  • Heavy metals (as Pb): ≤ 0.002 %
  • Ca (Calcium): ≤ 0.005 %
  • Fe (Iron): ≤ 0.001 %
  • K (Potassium): ≤ 0.005 %
  • Mg (Magnesium): ≤ 0.001 %
  • Na (Sodium): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service