Skip to Content
Merck
All Photos(1)

Key Documents

05-861

Sigma-Aldrich

Anti-vWF (von Willebrand Factor) Antibody, clone GMA-022

clone GMA-022, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

GMA-022, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

von Willebrand Factor

Immunogen

Purified human von Willebrand Factor

Application

Anti-vWF (von Willebrand Factor) Antibody, clone GMA-022 is an antibody against vWF (von Willebrand Factor) for use in WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Routinely evaluated by immunoblot

Target description

260kDa

Physical form

DEAE purified
Format: Purified

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shear stress and von Willebrand factor in health and disease.
Tsai, Han-Mou
Seminars in Thrombosis and Hemostasis, 29, 479-488 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service