890815P
Avanti
DOSPA
Avanti Research™ - A Croda Brand
Synonym(s):
2,3-dioleyloxy-N-[2-(sperminecarboxamido)ethyl]-N,N-dimethyl-1-propanaminium HCl salt
About This Item
Assay
>99% (TLC)
form
powder
packaging
pkg of 1x1 mg (890815P-1MG)
pkg of 1x5 mg (890815P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
application(s)
advanced drug delivery
lipid type
cationic lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
O=C(C([NH2+]CCC[NH3+])CCC[NH+]([H])CCC[NH3+])NCC[N+](C)(C)CC(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[5Cl-]
Packaging
5 mL Amber Glass Screw Cap Vial (890815P-5MG)
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service