Skip to Content
Merck
All Photos(2)

Key Documents

860372P

Avanti

16:0 PE-d62

Avanti Research - A Croda Brand 860372P, powder

Synonym(s):

1,2-dipalmitoyl-d62-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H12NO8PD62
CAS Number:
Molecular Weight:
754.34
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860372P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860372P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(C([2H])([2H]

General description

1,2-dipalmitoyl-d62-sn-glycero-3-phosphoethanolamine (16:0 PE-d62) is a deuterated phosphoethanolamine (PE) in which 62 protons of dipalmitoyl are replaced.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

1,2-dipalmitoyl-d62-sn-glycero-3-phosphoethanolamine (16:0 PE-d62) has been used as an apolar lipid internal standard for mass spectrometry. It may be used in the preparation of levuglandin hydroxylactam.

Packaging

5 mL Amber Glass Screw Cap Vial (860372P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Free radical biology & medicine, 47(11), 1539-1552 (2009-09-16)
Levuglandins (LGs) and isolevuglandins (isoLGs, also called "isoketals" or "isoKs") are extraordinarily reactive products of cyclooxygenase- and free radical-induced oxidation of arachidonates. We now report the detection in vivo and quantitative analysis of LG/isoLG adducts that incorporate the amino group
Xiyuan Lu et al.
Scientific reports, 7(1), 3455-3455 (2017-06-16)
To maintain core body temperature in cold conditions, mammals activate a complex multi-organ metabolic response for heat production. White adipose tissue (WAT) primarily functions as an energy reservoir, while brown adipose tissue (BAT) is activated during cold exposure to generate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service