Skip to Content
Merck
All Photos(1)

Key Documents

810197C

Avanti

16:0-Pyr PC

Avanti Research - A Croda Brand 810197C

Synonym(s):

1-hexadecanoyl-2-(1-pyrenedecanoyl)-sn-glycero-3-phosphatidylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H76NO8P
CAS Number:
Molecular Weight:
850.11
MDL number:
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810197C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810197C

concentration

1 mg/mL (810197C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCc1c2c3c4c(cc2)cccc4ccc3cc1)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C50H76NO8P/c1-5-6-7-8-9-10-11-12-13-14-17-20-23-29-47(52)56-39-45(40-58-60(54,55)57-38-37-51(2,3)4)59-48(53)30-24-21-18-15-16-19-22-26-41-31-32-44-34-33-42-27-25-28-43-35-36-46(41)50(44)49(42)43/h25,27-28,31-36,45H,5-24,26,29-30,37-40H2,1-4H3/t45-/m1/s1

InChI key

ZZLOWUUVDRBGKM-WBVITSLISA-N

General description

Phosphatidylcholine (PC) is a prime metabolite in eukaryotes glycerolipid metabolism, mainly synthesized in liver. It is a major element of lipoproteins, bile and lung surfactant.

Packaging

5 mL Amber Glass Screw Cap Vial (810197C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane activity of two short Trp-rich amphipathic peptides
Bozelli JC, et al.
Biochimica et biophysica acta. Proteins and proteomics null
Phosphatidylcholine biosynthesis and lipoprotein metabolism
Cole LK, et al.
Biochimica et Biophysica Acta null
Importance of phosphatidylcholine on the chloroplast surface
Botella C, et al.
Progress in Lipid Research null
Laura K Cole et al.
Biochimica et biophysica acta, 1821(5), 754-761 (2011-10-08)
Phosphatidylcholine (PC) is the major phospholipid component of all plasma lipoprotein classes. PC is the only phospholipid which is currently known to be required for lipoprotein assembly and secretion. Impaired hepatic PC biosynthesis significantly reduces the levels of circulating very
José C Bozelli et al.
Biochimica et biophysica acta. Biomembranes, 1862(7), 183280-183280 (2020-03-30)
Short linear antimicrobial peptides are attractive templates for developing new antibiotics. Here, it is described a study of the interaction between two short Trp-rich peptides, horine and verine-L, and model membranes. Isothermal titration calorimetry studies showed that the affinity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service