Key Documents
Z740725
Sigma-Aldrich
Aldrich® Essentials boiling flask, flat-bottom, ST/NS 24/40
capacity 1000 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
flat bottom
single-neck
packaging
pkg of 1 ea
capacity
1000 mL
Related Categories
General description
Clear borosilicate glass, flat bottomed boiling flasks with top, outer ST ground joint neck. White marking spot and capacity.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)