Z723169
Aldrich® Rotary Evaporator Adapter Set
joint: ST/NS 24/40
All Photos(2)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Recommended Products
Joint
joint: ST/NS24/40
Related Categories
General description
This versatile adapter connects to jars and bottles and to the vapor tube of a rotary evaporator. Precision machined from durable, chemically inert glass-filled PTFE. The unique, "universal fit" threaded connection adapts to any suitable cap.
Simply insert the threaded shaft of the adapter through a 16 mm diameter holed cap and secure by hand-tightening the PTFE gasket and nut. Attach the other end of the adapter to the vapor tube via standard taper joint and clip.
Field Tested: This adapter has been used extensively for evaporating solvents from asphalt and petroleum fractions using a standard size jar instead of a round-bottom flask from which sample recovery can be difficult. The adapter will be equally convenient for the recovery of solid materials in laboratory applications.
Adapter set includes: One 58 x 400 mm diameter holed cap and a solid top cap with PTFE liners, and a 200 mL borosilicate glass jar.
Simply insert the threaded shaft of the adapter through a 16 mm diameter holed cap and secure by hand-tightening the PTFE gasket and nut. Attach the other end of the adapter to the vapor tube via standard taper joint and clip.
Field Tested: This adapter has been used extensively for evaporating solvents from asphalt and petroleum fractions using a standard size jar instead of a round-bottom flask from which sample recovery can be difficult. The adapter will be equally convenient for the recovery of solid materials in laboratory applications.
Adapter set includes: One 58 x 400 mm diameter holed cap and a solid top cap with PTFE liners, and a 200 mL borosilicate glass jar.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)