Z676594
ART® low retention barrier tips
Tip design ART 20E, Reload insert
Synonym(s):
pipet tips, pipette tips
About This Item
RNase free
barrier
Reload insert
Tip design ART 20E
Recommended Products
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier
Reload insert
Tip design ART 20E
packaging
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename
Molecular BioProducts 2149E-05-RI
technique(s)
PCR: suitable
maximum volume
20 μL
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service