Z510815
Connecting Adapter
female joint: ST/NS 14/20, thread size 20-400
Synonym(s):
747130-2014
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Recommended Products
thread size
20-400
Joint
female joint: ST/NS14/20
General description
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Features and Benefits
- Accessory for microscale MICROFLEX threaded kits
- Connects Standard Taper outer ground joint to GPI thread of inner ground joint
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Choose from one of the most recent versions:
Certificates of Analysis (COA)