Key Documents
Z258458
Sigma-Aldrich
Head set for Vortex-Genie® 2
Large sample
Synonym(s):
Head Set for Vortex, Vortex Adapter, Vortex-Genie Head Set
About This Item
UNSPSC Code:
41103814
NACRES:
NB.73
manufacturer/tradename
Scientific Industries
General description
For continuous vortexing of beakers, flasks, and tubes. Includes a recessed platform, elastic flask harness, and an assortment of tube inserts (accepts 10 to 37mm diam. tubes).
Legal Information
Vortex-Genie is a registered trademark of Scientific Industries