Key Documents
Z189812
Sigma-Aldrich
Scienceware® industrial and lab funnel
white polyethylene, pkg of 1 ea
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
white polyethylene
grade
industrial grade
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H14716-0000
funnel H × O.D.
13 cm × 36 cm
stem H × O.D.
14 cm × 5.1 cm
General description
Molded, 1/4 in. PE funnel provides a greater range of chemical resistance. Fits any 2 in. drum opening.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP