Skip to Content
Merck
All Photos(1)

Documents

W218804

Sigma-Aldrich

Butyl isobutyrate

≥97%, FCC, FG

Synonym(s):

Butyl 2-methylpropanoate, Butyl isobutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2(CH2)3CH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2188
EC Number:
Council of Europe no.:
291
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.416
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.401 (lit.)

bp

155-156 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; banana; green; fruity; sweet

SMILES string

CCCCOC(=O)C(C)C

InChI

1S/C8H16O2/c1-4-5-6-10-8(9)7(2)3/h7H,4-6H2,1-3H3

InChI key

JSLCOZYBKYHZNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl isobutyrate is an aliphatic ester that can be used as a food flavoring agent. It is one of the main volatile flavor compounds present in Chardonnay musts.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of an aroma similar to that of sparkling wine: Sensory and gas chromatography analyses of fermented grape musts
Mamede ME, et al
Food Chemistry, 89(1), 63-68 (2005)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 345-346 (1997)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 345-346 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service