SYNC378040
Synthware™ chromatography column with reservoir and PTFE stopcock
1000 mL, top joint: ST/NS 24/40, I.D. × L 73.0 mm × 457 mm
Synonym(s):
Synthware C378040, glass chromatograpy column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS top24/40
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C378040
technique(s)
HPLC: suitable
I.D. × L
73.0 mm × 457 mm
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With 24/40 outer joint at top. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service