Skip to Content
Merck
All Photos(1)

Key Documents

H59206

Sigma-Aldrich

6-Hydroxy-2,4,5-triaminopyrimidine sulfate salt

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N5O · H2SO4
CAS Number:
Molecular Weight:
239.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

powder

SMILES string

Nc1nc(N)c(N)c(O)n1

InChI

1S/C4H7N5O/c5-1-2(6)8-4(7)9-3(1)10/h5H2,(H5,6,7,8,9,10)

InChI key

SYEYEGBZVSWYPK-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérôme Mulhbacher et al.
PLoS pathogens, 6(4), e1000865-e1000865 (2010-04-28)
Riboswitches are regulatory elements modulating gene expression in response to specific metabolite binding. It has been recently reported that riboswitch agonists may exhibit antimicrobial properties by binding to the riboswitch domain. Guanine riboswitches are involved in the regulation of transport

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service