Skip to Content
Merck
All Photos(1)

Key Documents

GF67677980

Carbon - vitreous - 3000c

granule, 2-3 mm nominal granule size, weight 25 g, maximum temperature 3000c in vacuum or inert gas (500-600c in air)

Synonym(s):

VC006303

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

granular

manufacturer/tradename

Goodfellow 676-779-80

weight

25 g

nominal granule size

2-3 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service