GF46333133
Constantan - resistance alloy, Cu55Ni45
insulated wire, 100m length, 0.05mm conductor diameter, 0.005mm insulation thickness, polyimide insulation
Synonym(s):
CU045820
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
polyimide insulation
thermocouple wire
form
wire (insulated)
manufacturer/tradename
Goodfellow 463-331-33
insulation thickness
0.005 mm
wire L × diam.
100 m × 0.05 mm , conductor diameter
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Application
may be used as one component of a thermocouple
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service