All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
Beilstein:
115875
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.579 (lit.)
bp
276 °C (lit.)
mp
8-10 °C (lit.)
density
1.054 g/mL at 25 °C (lit.)
SMILES string
C(c1ccccc1)c2ccccn2
InChI
1S/C12H11N/c1-2-6-11(7-3-1)10-12-8-4-5-9-13-12/h1-9H,10H2
InChI key
PCFUWBOSXMKGIP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
257.0 °F - closed cup
Flash Point(C)
125 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Angelica E Graminha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(4), 1073-1076 (2007-08-21)
Reaction of [RuCl(3)(dppb)H(2)O] (dppb=1,4 bis(diphenylphospine)butane) with 2-benzoylpyridine thiosemicarbazone (H2Bz4DH) and its N(4)-methyl (H2Bz4M) and N(4)-phehyl (H2Bz4Ph) derivatives gave [RuCl(dppb)(H2Bz4DH)]Cl (1), [RuCl(dppb)(H2Bz4M)]Cl (2) and [RuCl(dppb)(H2Bz4Ph)]Cl (3). The cytotoxic activity of the studied compounds was tested against the MCF-7, TK-10 and UACC-62
Masaki Itoh et al.
Organic letters, 16(7), 2050-2053 (2014-03-22)
The direct α-methylenation of benzylpyridines was achieved using N,N-dimethylacetamide (DMA) as a one-carbon source under copper catalysis. An intermediary species was detected at an early stage, and a possible mechanism was proposed. Additionally, α-oxygenation and dimerization of benzylpyridines could also
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service