Skip to Content
Merck
All Photos(1)

Key Documents

928380

Sigma-Aldrich

5-{4-[2-(2-{[(tert-butoxy)carbonyl]amino}ethoxy)ethyl]piperazin-1-yl}pyrimidine-2-carboxylic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29N5O5
Molecular Weight:
395.45
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: linker

functional group

Boc
amine
carboxylic acid

storage temp.

2-8°C

Application

5-{4-[2-(2-{[(tert-butoxy)carbonyl]amino}ethoxy)ethyl]piperazin-1-yl}pyrimidine-2-carboxylic acid is a heterobifunctional crosslinker useful for development of PROTAC degraders for targeted protein degradation or other bifunctional molecules in chemical biology. Incorporation of rigidity into the linker region may impact the 3D orientation of the molecules and thus ternary complex formation as well as optimization of drug-like properties. For linkers already conjugated to E3 ligase ligands, please visit our collection of Protein Degrader Building Blocks.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service