Skip to Content
Merck
All Photos(2)

Key Documents

900021

Sigma-Aldrich

TBAF(pin)2

Synonym(s):

1-Butanaminium, N,N,N-tributyl-, fluoride, compd. with 2,3-dimethyl-2,3-butanediol (1:2:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H64FNO4
CAS Number:
Molecular Weight:
497.81
UNSPSC Code:
12352101
PubChem Substance ID:

form

powder

mp

162-167 °C (d)

functional group

amine
hydroxyl

SMILES string

OC(C)(C)C(C)(C)O.OC(C)(C)C(C)(C)O.CCCC[N+](CCCC)(CCCC)CCCC.[F-]

InChI

1S/C16H36N.2C6H14O2.FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2*1-5(2,7)6(3,4)8;/h5-16H2,1-4H3;2*7-8H,1-4H3;1H/q+1;;;/p-1

InChI key

CVEUWEZBMVLEKR-UHFFFAOYSA-M

General description

TBAF(pin)2 is a fluoride–alcohol complex with coordination number four. It can be obtained by reacting pinacol (pin) with tetrabutylammonium fluoride trihydrate (TBAF(H2O)3). The geometric structure and bonding of TBAF(pin)2 have been studied.

Application

Nucleophilic fluorinating reagent is a more convenient crystalline solid for handling and storage compared to tetrabutylammonium fluoride hydrate (Aldrich 241512). As reported by Engle and Gouverneur, these solids display comparable reactivity on similar substrates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coordination diversity in hydrogen-bonded homoleptic fluoride?alcohol complexes modulates reactivity.
Engle KM, et al.
Chemical Science, 6(9), 5293-5302 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service