Skip to Content
Merck
All Photos(1)

Key Documents

774545

Sigma-Aldrich

Nickel(II) oxide

nanowires, diam. × L ~20 nm × 10 μm

Synonym(s):

Nickel Monoxide, Nickel oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiO
CAS Number:
Molecular Weight:
74.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanowires

Quality Level

diam. × L

~20 nm × 10 μm

color

black

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nickel oxide nanowires have been studied for use in magnetic memory devices; as well as in luminescent materials.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Oka et al.
Journal of the American Chemical Society, 131(10), 3434-3435 (2009-02-21)
We have demonstrated the nonvolatile bipolar resistive memory switching in single crystalline NiO heterostructured nanowires for the first time. The self-assembled NiO nanowires are expected to open up opportunities to explore not only the detailed nanoscale mechanisms in NiO resistive
Lei Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 34(31), 9314-9321 (2018-07-13)
Selective extraction of a small amount of nucleic acids from complex biological samples containing a high concentration of proteins is critical for bioanalytical chemistry. A number of previously published studies have focused on long, double-stranded DNA such as plasmid DNA.
Z Y Wu et al.
The journal of physical chemistry. B, 109(7), 2512-2515 (2006-07-21)
Nickel oxide nanowires modified by poly(vinylpyrrolidone) (PVP) were synthesized via a simple chemical pattern. For the first time NiO nanowires with diameters ranging from 40 to 100 nm with the expected ratio (length vs diameter) ranging from 54 to 90
Milena E Teixeira et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(8), 773-780 (2015-08-11)
In this work the electrooxidations of glucose, galactose, mannose, rhamnose, xylose and arabinose are studied at a nickel hydroxide nanoparticle modified boron-doped diamond electrode and compared to an unmodified electrode. These carbohydrates are very important in the second-generation ethanol production
Marcia Regina Salvadori et al.
PloS one, 10(6), e0129799-e0129799 (2015-06-05)
The use of dead biomass of the fungus Hypocrea lixii as a biological system is a new, effective and environmentally friendly bioprocess for the production and uptake of nickel oxide nanoparticles (NPs), which has become a promising field in nanobiotechnology.

Articles

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service