Skip to Content
Merck
All Photos(1)

Key Documents

737240

Sigma-Aldrich

(1-Bromo-2,2,2-trifluoroethyl)benzene

95%

Synonym(s):

(1-Bromotrifluoroethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrF3
CAS Number:
Molecular Weight:
239.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.487

density

1.555 g/mL at 25 °C

functional group

bromo
fluoro
phenyl

SMILES string

FC(F)(F)C(Br)c1ccccc1

InChI

1S/C8H6BrF3/c9-7(8(10,11)12)6-4-2-1-3-5-6/h1-5,7H

InChI key

IRICHAOGAOFEQI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

164.0 °F

Flash Point(C)

73.33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service