728276
(Bromoethynyl)benzene
≥95.0% (HPLC)
Synonym(s):
1-Bromo-2-phenylacetylene, 1-Bromo-2-phenylethyne, Phenylethynyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H5Br
CAS Number:
Molecular Weight:
181.03
Beilstein:
606222
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0% (HPLC)
form
liquid
storage temp.
−20°C
SMILES string
BrC#Cc1ccccc1
InChI
1S/C8H5Br/c9-7-6-8-4-2-1-3-5-8/h1-5H
InChI key
BPVHWNVBBDHIQU-UHFFFAOYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service