Skip to Content
Merck
All Photos(1)

Key Documents

727431

Sigma-Aldrich

4-Azidoanisole solution

~0.5 M in tert-butyl methyl ether, ≥90.0% (HPLC)

Synonym(s):

1-Azido-4-methoxybenzene solution, 4-Methoxyphenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3O
CAS Number:
Molecular Weight:
149.15
Beilstein:
3609304
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥90.0% (HPLC)

Quality Level

Assay

≥90.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

COc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C7H7N3O/c1-11-7-4-2-6(3-5-7)9-10-8/h2-5H,1H3

InChI key

PPKDSHDYUBDVKL-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service