669474
2-Bromo-4′-(trifluoromethoxy)acetophenone
97%
Synonym(s):
4-(Trifluoromethoxy)phenacyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H6BrF3O2
CAS Number:
Molecular Weight:
283.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
48-55 °C
functional group
bromo
fluoro
ketone
storage temp.
2-8°C
SMILES string
FC(F)(F)Oc1ccc(cc1)C(=O)CBr
InChI
1S/C9H6BrF3O2/c10-5-8(14)6-1-3-7(4-2-6)15-9(11,12)13/h1-4H,5H2
InChI key
AOAGGWLQIILIIV-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service